Methyl Indole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 1H-indole-2-carboxylate, Indole-2-carboxylic Acid Methyl Ester
A187053-100G
A187053-10G
A187053-1G
A187053-25G
A187053-5G
103815-986EA
5
USD
103815-986
103846-574
76799-886
76799-888
77084-262
77084-264
77084-266
Methyl Indole-2-carboxylate 95%
Methyl Indole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 331.7 °C (760 mm Hg) Melting Pt: 151…155 °C Flash Pt: 154.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460779 CAS Number: 1202-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...