Methyl-1H-imidazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-imidazolecarboxylate, Methyl 1H-imidazole-5-carboxylate

A219749-1G A219749-25G A219749-5G A219749-100G A219749-10G
103818-810EA 5 USD
103818-810 103849-690 77088-246 77088-248 77088-250 77097-940 77097-942
Methyl-1H-imidazole-5-carboxylate 98%
Methyl-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Boiling Pt: 318.2 °C (760 mm Hg)
Melting Pt: 154…156 °C
Flash Pt: 146.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00216584
CAS Number: 17325-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR