Methyl-1H-imidazole-2-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 1H-imidazole-2-carboxylate

A128109-100G A128109-10G A128109-1G A128109-25G A128109-5G
103801-072EA 8 USD
103801-072 76811-028 76811-030 77313-358 77313-360 77313-362
Methyl-1H-imidazole-2-carboxylate 96%
Methyl-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.1100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203842
CAS Number: 17334-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR