Methyl-1-cyclopentene-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-cyclopentene-1-carboxylate

A150792-100G A150792-10G A150792-1G A150792-5G
103804-200EA 6 USD
103804-200 103842-338 77095-802 77095-804 77317-866 77317-868
Methyl-1-cyclopentene-1-carboxylate 95%
Methyl 1-cyclopentene-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.1531 g/mol
Boiling Pt: 161.3 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00239506
CAS Number: 25662-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR