Methyl-1-boc-3-pyrrolidinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate

A102875-100G A102875-10G A102875-25G A102875-1G A102875-5G
103796-434EA 10 USD
103796-434 103822-134 77107-292 77107-294 77107-296 77309-002 77309-004
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Methyl-1-boc-3-pyrrolidinecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Flash Pt: 128.32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038683
CAS Number: 122684-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR