Methyl-1-benzylpiperidine-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-benzylpiperidine-4-carboxylate

A249931-100G A249931-25G A249931-5G
103851-742EA 6 USD
103851-742 77031-478 77091-090 77091-092
Methyl-1-benzylpiperidine-4-carboxylate 95%
Methyl-1-benzylpiperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.3100 g/mol
Boiling Pt: 310.7 °C (760 mm Hg)
Flash Pt: 108.437 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750949
CAS Number: 10315-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR