Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A123040-10G
A123040-1G
A123040-500G
A123040-5G
A123040-100G
A123040-25G
76766-674EA
12
USD
76766-674
76766-676
76766-678
77312-452
77624-024
77624-026
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95%
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl Boiling Pt: 366 °C (760 mm Hg) Melting Pt: 185 °C Flash Pt: 175.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012799 CAS Number: 3939-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...