Methyl-1-aminocyclopropanecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Aminocyclopropane-1-carboxylic acid Methyl Ester Hydrochloride, 1-Aminocyclopropanecarboxylic acid methyll ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-Aminocyclopropanecarboxylic acid methyl ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate HCl

A194658-1G A194658-100G A194658-250MG A194658-5G A194658-10G
103816-680EA 9 USD
103816-680 103847-290 77085-214 77192-100 77192-102 77302-426 77615-642
Methyl-1-aminocyclopropanecarboxylate hydrochloride 97%
Methyl-1-aminocyclopropanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.5914 g/mol
Melting Pt: 185…186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078879
CAS Number: 72784-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR