Methyl-1-aminocyclopropane-1-carboxylate 97%

Supplier: Ambeed
Warning

A283590-1G A283590-10G
103813-620EA 10 USD
103813-620 103853-570 76837-282 77872-359
Methyl-1-aminocyclopropane-1-carboxylate 97%
Methyl-1-aminocyclopropane-1-carboxylate

Precaución: for Research Use Only

Formula: C₅H₉NO₂
Boiling Pt: 136.4 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00906963
CAS Number: 72784-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR