Methyl 1-amino-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

A161098-100MG A161098-250MG A161098-1G
103805-450EA 198 USD
103805-450 103843-688 76872-016 76872-018 77300-514
Methyl 1-amino-1H-pyrrole-2-carboxylate 95%
Methyl 1-amino-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 272.3 °C (760 mm Hg)
Flash Pt: 118.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11226170
CAS Number: 122181-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR