Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1,4-dioxaspiro[4.5]decane-8-carboxylate, 1,4-Dioxaspiro[4.5]decane-8-carboxylic acid methyl ester

A159513-1G A159513-25G A159513-5G
103805-260EA 19 USD
103805-260 103843-480 77319-368 77319-370 77319-372
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 97%
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195278
CAS Number: 26845-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR