Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A108948-250MG A108948-1G
103797-410EA 39 USD
103797-410 103823-200 76982-608 77310-030
Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate 95%
Methyl 1,3-dimethyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Boiling Pt: 242.0±20.0 °C (760 mm Hg)
Flash Pt: 100.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06204177
CAS Number: 10250-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR