Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate

A101119-10G A101119-1G A101119-250MG
103795-810EA 40 USD
103795-810 103821-500 76780-464 77296-990 77666-462
Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate 98%
Methyl-1-(5-bromopyrimidin-2-yl)piperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN₃O₂
MW: 300.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275691
CAS Number: 914347-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR