1-tert-Butyl 2-methyl (2R)-piperazine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butyl) 2-methyl (R)-piperazine-1,2-dicarboxylate, Methyl (R)-1-Boc-piperazine-2-carboxylate

A132307-1G A132307-5G
103801-714EA 46 USD
103801-714 77314-288 77314-290
1-tert-Butyl 2-methyl (2R)-piperazine-1,2-dicarboxylate 97%
1-tert-Butyl 2-methyl (2R)-piperazine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.287 g/mol
Boiling Pt: 321.3 °C (760 mm Hg)
Flash Pt: 148.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115327
CAS Number: 252990-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR