(R)-(-)-Methyl mandelate 97%

Supplier: Ambeed
Warning

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

A183926-100G A183926-10G A183926-1G A183926-25G A183926-5G
103815-688EA 8 USD
103815-688 103846-250 77248-364 77248-366 77301-864 77301-866 77301-868
(R)-(-)-Methyl mandelate 97%
(R)-(-)-Methyl mandelate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 258.1 °C (760 mm Hg)
Melting Pt: 56…58 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00064247
CAS Number: 20698-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR