Fenbendazole 98+%
Supplier: Ambeed
|
Danger
|
A167022-100G
A167022-100MG
A167022-500G
A167022-1G
A167022-25G
A167022-5G
A167022-10G
103806-122EA
9
USD
103806-122
103844-384
76799-238
76799-240
76799-242
77300-868
77300-870
77300-872
77879-117
Fenbendazole 98+%
Fenbendazole
Precaución: for Research Use Only
Formula:
C₁₅H₁₃N₃O₂S Melting Pt: 233 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144301 CAS Number: 43210-67-9 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...