(4-Cyanophenyl)acetic acid methyl ester 96%

Supplier: Ambeed
Warning

Synonyms: 4-Cyanobenzeneacetic acid methyl ester, Methyl (4-cyanophenyl)acetate

A382231-5G A382231-250MG A382231-1G A382231-25G A382231-100G A382231-10G
103826-710EA 16 USD
103826-710 103837-746 76841-066 77168-080 77306-930 77306-932 77684-102 77684-103
(4-Cyanophenyl)acetic acid methyl ester 96%
(4-Cyanophenyl)acetic acid methyl ester

Precaución: For research use only.

Formula: C₁₀H₉NO₂
MW: 175.1840 g/mol
Boiling Pt: 289.9 °C (760 mm Hg)
Melting Pt: 40…60 °C
Flash Pt: 132.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02181140
CAS Number: 52798-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR