Boc-cis-4-amino-L-proline methyl ester hydrochloride salt 97%

Supplier: Ambeed
Warning

A306464-100MG A306464-250MG A306464-1G A306464-5G
103814-468EA 36 USD
103814-468 103835-634 76723-732 76723-734 76723-736 76838-344 76838-346
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt 97%
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt

Precaución: for Research Use Only

Formula: C₁₁H₂₁ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD03787927
CAS Number: 171110-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR