Methocarbamol 98%

Supplier: Ambeed
Warning

A160451-100G A160451-100MG A160451-10G A160451-25MG A160451-500G A160451-25G A160451-1MG
103843-610EA 12 USD
103843-610 76798-988 76798-990 76798-992 76798-994 76798-996 77319-546 77667-250
Methocarbamol 98%
Methocarbamol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₅
Boiling Pt: 472.5 °C (760 mm Hg)
Melting Pt: 95…97 °C
Flash Pt: 239.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00057662
CAS Number: 532-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR