1,1-Dimethylbiguanide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride
A122997-100G
A122997-10G
A122997-25G
A122997-500G
A122997-5G
103808-262EA
78
USD
103808-262
77108-666
77108-668
77108-670
77108-672
77297-914
1,1-Dimethylbiguanide hydrochloride 97%
1,1-Dimethylbiguanide hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₂ClN₅ Boiling Pt: 224.1 °C (760 mm Hg) Melting Pt: 223…226 °C Flash Pt: 89.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012582 CAS Number: 1115-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...