1-Mesitylhydrazine hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: Mesitylhydrazine hydrochloride

A153688-250MG A153688-1G A153688-25G A153688-5G
103804-552EA 22 USD
103804-552 103842-726 76871-744 77318-400 77318-402 77318-404
1-Mesitylhydrazine hydrochloride 95%
1-Mesitylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄N₂·HCl
MW: 186.69 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00052268
CAS Number: 24006-09-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR