Hydroquinone monomethyl ether 98%

Supplier: Ambeed
Warning

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

A245207-25G A245207-1000G A245207-100G A245207-10G A245207-500G
103851-462EA 6 USD
103851-462 77090-702 77288-844 77288-846 77288-848 77288-850
Hydroquinone monomethyl ether 98%
Hydroquinone monomethyl ether

Precaución: for Research Use Only

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 243
Melting Pt: 56 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR