Melatonin 97%

Supplier: Ambeed
Warning

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

A204285-100G A204285-10G A204285-500G
103848-200EA 9 USD
103848-200 76721-514 77667-704 77667-706
Melatonin 97%
Melatonin

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Boiling Pt: 512.8 °C (760 mm Hg)
Melting Pt: 116.5…118 °C
Flash Pt: 264 °C
Storage Temperature: Freezer
MDL Number: MFCD00005655
CAS Number: 73-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR