Mebendazole 97%

Supplier: Ambeed
Warning

Synonyms: Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate, Methyl (5-benzoyl-1H-benzo[d]imidazol-2-yl)carbamate

A162607-100G A162607-10G A162607-500G A162607-5G A162607-1000G
103843-886EA 6 USD
103843-886 76768-634 76768-636 76768-638 77300-600 77625-584
Mebendazole 97%
Mebendazole

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃O₃
MW: 295.30 g/mol
Melting Pt: 288.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00057872
CAS Number: 31431-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR