Manganese(III) acetate dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: Acetic acid manganese (III) salt dihydrate, Manganese triacetate dihydrate

A185213-100G A185213-10G A185213-25G A185213-5G
103846-396EA 14 USD
103846-396 76799-836 76799-838 77084-028 77084-030
Manganese(III) acetate dihydrate 98%
Manganese(III) acetate dihydrate

Precaución: for Research Use Only

Formula: (H₃CCOO)₂Mn·2H₂O
MW: 209.06
Storage Temperature: Ambient
MDL Number: MFCD00150022
CAS Number: 19513-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR