2-Morpholino-8-phenylchromone 98%

Supplier: Ambeed
Warning

Synonyms: LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran

A133122-1MG A133122-10MG A133122-50MG A133122-100MG A133122-250MG
103792-136EA 40 USD
103792-136 103792-138 103799-348 103801-830 76797-954 77314-438 77314-440 77666-908 77666-910
2-Morpholino-8-phenylchromone 98%
2-Morpholino-8-phenylchromone

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₃
MW: 307.35 g/mol
Boiling Pt: 494.6 °C (760 mm Hg)
Melting Pt: 183…185 °C
Flash Pt: 253 °C
Storage Temperature: Freezer
MDL Number: MFCD00270881
CAS Number: 154447-36-6
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR