(S)-(+)-2-Amino-3-methyl-1-butanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol
A200742-100G
A200742-10G
A200742-500G
A200742-25G
A200742-5G
A200742-1G
103817-234EA
5
USD
103817-234
103847-868
76986-454
76986-456
76986-458
77085-930
77085-932
77302-716
(S)-(+)-2-Amino-3-methyl-1-butanol 97%
(S)-(+)-2-Amino-3-methyl-1-butanol
Precaución: for Research Use Only
Formula:
C₅H₁₃NO MW: 103.16 g/mol Boiling Pt: 186.8 °C (760 mm Hg) Melting Pt: 30…34 °C Flash Pt: 78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064296 CAS Number: 2026-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...