Methyl-L-threoninate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-Methyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OMe·HCl, L-Threonine methyl ester hydrochloride

A250411-100G A250411-10G A250411-25G A250411-5G A250411-1G
103811-922EA 9 USD
103811-922 103851-762 76987-870 76987-872 77091-112 77091-114 77304-526
Methyl-L-threoninate hydrochloride 97%
Methyl-L-threoninate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₃
Melting Pt: 64 °C
Flash Pt: 119.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00037677
CAS Number: 39994-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR