L-Theanine 98%

Supplier: Ambeed
Warning

A320722-25G A320722-100G A320722-100MG A320722-10G A320722-1MG A320722-25MG A320722-500G
103836-178EA 15 USD
103836-178 76839-060 77114-716 77114-718 77114-720 77114-722 77114-724 77612-854
L-Theanine 98%
L-Theanine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₃
Boiling Pt: 430.2 °C (760 mm Hg)
Melting Pt: 270 °C
Flash Pt: 214 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059653
CAS Number: 3081-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR