L-tert-Leucinol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Leucinol, (S)-2-Amino-3,3-dimethylbutan-1-ol, (S)-2-Amino-3,3-dimethylbutanol

A342037-1G A342037-25G A342037-5G A342037-100G A342037-10G A342037-500G
103825-856EA 31 USD
103825-856 103836-830 76839-862 76839-864 76839-866 77003-156 77003-158 77003-160
L-tert-Leucinol 98%
L-tert-Leucinol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 70 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192250
CAS Number: 112245-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR