L(+)-Rhamnose monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate

A827084-1000G A827084-100G A827084-10G A827084-500G
103841-160EA 5 USD
103841-160 76776-624 76776-626 76776-628 76776-630
L(+)-Rhamnose monohydrate 98%
L(+)-Rhamnose monohydrate

Precaución: for Research Use Only

Formula: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Boiling Pt: 323.9 °C (760 mm Hg)
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0
Merck Index: 12,08338

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR