Losartan 98%

Supplier: Ambeed
Danger

A254713-100G A254713-10G A254713-1MG A254713-250MG A254713-25G A254713-5G
103812-156EA 8 USD
103812-156 76801-648 76801-650 76801-652 76801-654 76801-656 76801-658
Losartan 98%
Losartan

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClN₆O
Boiling Pt: 682 °C (760 mm Hg)
Melting Pt: 183…184 °C
Flash Pt: 366.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865831
CAS Number: 114798-26-4
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR