Loratadine 98%

Supplier: Ambeed
Warning

A188364-100G A188364-10G A188364-1G A188364-25G A188364-5G A188364-100MG
103816-092EA 7 USD
103816-092 103846-684 76872-986 76872-988 77302-120 77302-122 77302-124 77883-400
Loratadine 98%
Loratadine

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClN₂O₂
Boiling Pt: 531.3 °C (760 mm Hg)
Melting Pt: 134…136 °C
Flash Pt: 275.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672869
CAS Number: 79794-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR