Lomefloxacin hydrochloride 95%

Supplier: Ambeed
Warning

A215886-25MG A215886-5G A215886-25G A215886-1G
103849-234EA 24 USD
103849-234 76800-758 77087-796 77667-812 77879-326
Lomefloxacin hydrochloride 95%
Lomefloxacin hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₉F₂N₃O₃·HCl
Melting Pt: 290…300 °C
Flash Pt: 282 °C
Storage Temperature: Freezer
MDL Number: MFCD00214312
CAS Number: 98079-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR