Loganin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,4aS,6S,7R,7aS)-Methyl 6-hydroxy-7-methyl-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate

A106244-1MG A106244-250MG A106244-50MG A106244-5MG A106244-10MG A106244-100MG
103791-418EA 13 USD
103791-418 103791-984 77107-664 77107-666 77107-668 77107-670 77283-452 77684-965
Loganin 98%
Loganin

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₁₀
MW: 390.39 g/mol
Boiling Pt: 608.8 °C (760 mm Hg)
Melting Pt: 222 °C
Flash Pt: 217.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00075645
CAS Number: 18524-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR