N,N-Diethyl-4-(4-((3-(piperidin-1-yl)propyl)amino)quinolin-2-yl)benzamide dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: LMPTP inhibitor 1 2HCl, LMPTP Inhibitor 1 dihydrochloride

A564234-100MG A564234-1MG A564234-10MG A564234-25MG A564234-5MG A564234-50MG
103791-694EA 93 USD
103791-694 103792-770 103792-772 103799-684 76649-832 76649-834 76844-018 76844-020 76844-022 77307-818
N,N-Diethyl-4-(4-((3-(piperidin-1-yl)propyl)amino)quinolin-2-yl)benzamide dihydrochloride 97%
N,N-Diethyl-4-(4-((3-(piperidin-1-yl)propyl)amino)quinolin-2-yl)benzamide dihydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₃₈Cl₂N₄O
MW: 517.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31807303
CAS Number: 2310135-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR