Lithium difluoro(oxalato)borate 95%
Supplier: Ambeed
|
Warning
|
A548767-1G
A548767-25G
A548767-5G
A548767-10G
103828-678EA
6
USD
103828-678
103839-800
76843-870
76843-872
76843-874
77169-702
Lithium difluoro(oxalato)borate 95%
Lithium difluoro(oxalato)borate
Precaución: for Research Use Only
Formula:
C₂BF₂LiO₄ MW: 143.76 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27952543 CAS Number: 409071-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...