L-Isoserine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3-Amino-2-hydroxypropanoic acid, L-Isoserine, (S)-3-Amino-2-hydroxypropionic Acid
A205814-10G
A205814-1G
A205814-25G
A205814-5G
A205814-100G
103817-664EA
10
USD
103817-664
103848-330
76975-336
77086-556
77086-558
77086-560
77612-016
L-Isoserine 97%
L-Isoserine
Precaución: for Research Use Only
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Boiling Pt: 386.6 °C (760 mm Hg) Melting Pt: 199…201 °C Flash Pt: 187.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03428326 CAS Number: 632-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...