L-(-)-Fucose 98%

Supplier: Ambeed
Warning

Synonyms: (-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal

A101796-10G A101796-250MG A101796-25G A101796-1G A101796-5G A101796-100G
103796-142EA 7 USD
103796-142 103821-830 76994-534 76994-536 76994-538 77308-726 77308-728 77548-918
L-(-)-Fucose 98%
L(-)-Fucose

Precaución: for Research Use Only

Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Boiling Pt: 323.9 °C (760 mm Hg)
Melting Pt: 150…153 °C
Flash Pt: 149.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00069812
CAS Number: 2438-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR