Levetiracetam 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-Oxopyrrolidin-1-yl)butanamide, (S)-2-(2-Oxo-1-pyrrolidinyl)butyramide

A185961-100G A185961-10G A185961-5G A185961-1G
103815-898EA 11 USD
103815-898 103846-472 77029-620 77029-622 77084-122 77302-004
Levetiracetam 98%
Levetiracetam

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
Melting Pt: 118…121 °C
Flash Pt: 193.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03265610
CAS Number: 102767-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR