L(-)-Carnitine 98%

Supplier: Ambeed
Warning

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

A374826-25G A374826-100G A374826-10G A374826-1MG A374826-500G
103810-236EA 225 USD
103810-236 103837-580 76840-864 77101-614 77101-616 77101-618 77101-620
L(-)-Carnitine 98%
L(-)-Carnitine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00038747
CAS Number: 541-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR