(S)-2-(3-Aminopropanamido)-3-(1-methyl-1H-imidazol-5-yl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Anserine, N-beta-Alanyl-3-methyl-L-histidine

A265984-100MG A265984-250MG A265984-1G A265984-25G A265984-5G
103812-768EA 16 USD
103812-768 103852-664 77014-300 77014-302 77304-936 77304-938 77304-940
(S)-2-(3-Aminopropanamido)-3-(1-methyl-1H-imidazol-5-yl)propanoic acid 97%
(S)-2-(3-Aminopropanamido)-3-(1-methyl-1H-imidazol-5-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄O₃
MW: 240.25 g/mol
Boiling Pt: 611.3 °C (760 mm Hg)
Flash Pt: 323.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11040910
CAS Number: 584-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR