Lafutidine ≥99%

Supplier: Ambeed
Warning

Synonyms: (+)-(Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, FRG-8813, (Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((3-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, (+)-Lafutidine

A101474-1G A101474-5G
103795-992EA 37 USD
103795-992 77666-464 77666-466
Lafutidine ≥99%
Lafutidine

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₂₂H₂₉N₃O₄S
MW: 431.55 g/mol
Boiling Pt: 704.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD21607521
CAS Number: 118288-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR