6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid, Isoxepac, 2-(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid

A276039-100G A276039-10G A276039-500G A276039-25G A276039-5G
103853-218EA 6 USD
103853-218 77032-052 77032-054 77032-056 77305-226 77305-228
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid 98%
6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₄
MW: 268.27 g/mol
Boiling Pt: 528.2 °C (760 mm Hg)
Melting Pt: 130…132 °C
Flash Pt: 203.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00242952
CAS Number: 55453-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR