5-Isoxazolecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isoxazole-5-carboxylic acid

A107152-10G A107152-250MG A107152-25G A107152-5G
103797-136EA 16 USD
103797-136 103822-896 76809-712 76809-714 76809-716 77309-760
5-Isoxazolecarboxylic acid 98%
5-Isoxazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃NO₃
MW: 113.07 g/mol
Boiling Pt: 321.2 °C (760 mm Hg)
Melting Pt: 144…148 °C
Flash Pt: 148.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00156151
CAS Number: 21169-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR