4-Isoxazolecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isoxazole-4-carboxylic acid
A169793-100MG
A169793-250MG
A169793-5G
103806-410EA
33
USD
103806-410
103844-674
77163-494
77163-496
77300-998
4-Isoxazolecarboxylic acid 98%
4-Isoxazolecarboxylic acid
Precaución: Keep in a dark place, sealed in dry, at 2…8°C
Formula:
C₄H₃NO₃ MW: 113.0715 g/mol Boiling Pt: 321.2 °C (760 mm Hg) Flash Pt: 148.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06738806 CAS Number: 6436-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...