Isoxazol-5-ylmethanamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A176688-100MG A176688-250MG A176688-1G A176688-50MG
103799-432EA 61 USD
103799-432 103807-162 77163-736 77163-738 77301-462 77301-464
Isoxazol-5-ylmethanamine hydrochloride ≥95%
Isoxazol-5-ylmethanamine hydrochloride

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06738817
CAS Number: 440099-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR