3-(Hydroxymethyl)isoxazole 98%

Supplier: Ambeed
Warning

Synonyms: Isoxazol-3-ylmethanol

A414908-5G A414908-10G A414908-250MG
103827-284EA 55 USD
103827-284 103838-328 76841-902 77115-868 77115-870
3-(Hydroxymethyl)isoxazole 98%
3-(Hydroxymethyl)isoxazole

Precaución: for Research Use Only

Formula: C₄H₅NO₂
MW: 99.088 g/mol
Boiling Pt: 240 °C (760 mm Hg)
Flash Pt: 98.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778413
CAS Number: 89102-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR