3-Aminoisoxazole 98%

Supplier: Ambeed
Warning

A206388-100G A206388-10G A206388-1G A206388-25G A206388-5G
103817-708EA 5 USD
103817-708 103848-380 76975-366 76975-368 77086-632 77086-634 77086-636
3-Aminoisoxazole 98%
3-Aminoisoxazole

Precaución: for Research Use Only

Formula: C₃H₄N₂O
MW: 84.08 g/mol
Boiling Pt: 234.7 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038814
CAS Number: 1750-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR