3-Hydroxy-p-anisic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-methoxybenzoic acid, Isovanillic acid

A132591-100G A132591-5G
76688-696EA 79.57 USD
76688-696 77298-564
3-Hydroxy-p-anisic acid 95%
3-Hydroxy-p-anisic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 344.3 °C (760 mm Hg)
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00002507
CAS Number: 645-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR